Proteintech
Manufacturer Code:126881AP150UL
Catalog # 50-172-8175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BHLHE41 Fusion Protein Ag3381 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix domain containing, class B3; basic helix-loop-helix family, member e41; bHLH transcriptional factor Dec2; bHLHb3; BHLHB3 BHLHE41 DEC2 hDEC2 SHARP 1 SHARP1; bHLHe41; Class B basic helix-loop-helix protein 3; Class E basic helix-loop-helix protein 41; Differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; mDEC2; SHARP-1
Gene Aliases: 6430520M22Rik; Bhlhb2l; BHLHB3; BHLHE41; DEC2; hDEC2; SHARP-1; SHARP1
UniProt ID: (Human) Q9C0J9, (Mouse) Q99PV5
Entrez Gene ID: (Human) 79365, (Rat) 117095, (Mouse) 79362
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.