Invitrogen
Manufacturer Code:PA564152
Catalog # PIPA564152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BHLHB5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix family, member e22; bHLHb5; BHLHB5 Beta3 CAGL85 TNRC20 basic helix-loop-helix domain containing class B 5 class B basic helix-loop-helix protein 5 class E basic helix-loop-helix protein 22 trinucleotide repeat containing 20 trinucleotide repeat-containing gene 20 protein; bHLHe22; Class B basic helix-loop-helix protein 5; Class E basic helix-loop-helix protein 22; trinucleotide repeat containing 20; Trinucleotide repeat-containing gene 20 protein
Gene Aliases: Beta3; BHLHB5; BHLHE22; CAGL85; TNRC20
UniProt ID: (Human) Q8NFJ8
Entrez Gene ID: (Human) 27319
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.