Invitrogen
Manufacturer Code:PA5100405
Catalog # PIPA5100405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BHLHE22 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix (bHLH) gene class B beta3 basic helix-loop-helix domain containing class B 5 basic helix-loop-helix domain containing class B5 basic helix-loop-helix family member e22 basic helix-loop-helix family member e22 Beta3 Bhlhb5 Bhlhe22 CAGL85 Class B basic helix-loop-helix protein 5 class E basic helix-loop-helix protein 22 Protein BETA3 TNRC20 trinucleotide repeat containing 20 trinucleotide repeat-containing gene 20 protein; basic helix-loop-helix (bHLH) gene, class B, beta3; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix domain containing, class B5; basic helix-loop-helix family, member e22; bHLHb5; bHLHe22; Class B basic helix-loop-helix protein 5; Class E basic helix-loop-helix protein 22; Protein BETA3; trinucleotide repeat containing 20; Trinucleotide repeat-containing gene 20 protein
Gene Aliases: Beta3; BHLHB5; BHLHE22; CAGL85; TNRC20
UniProt ID: (Human) Q8NFJ8, (Mouse) Q8C6A8
Entrez Gene ID: (Human) 27319, (Mouse) 59058, (Rat) 365748
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.