Novus Biologicals
Manufacturer Code:NBP239085
Catalog # NBP239085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: GLLVGSAGVGGDPSLSSLPAGAALCLKYGESASRGSVAESSGGEQSPDDDSDGRCELVLRAGV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic Helix-Loop-Helix Domain Containing Class B 5 basic helix-loop-helix family member e22 Beta3 BHLHE22 CAGL85 Class B Basic Helix-Loop-Helix Protein 5 Class E Basic Helix-Loop-Helix Protein 22 TNRC20 Trinucleotide Repeat Containing 20 Trinucleotide Repeat-Containing Gene 20 Protein; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix family, member e22; bHLHb5; bHLHe22; Class B basic helix-loop-helix protein 5; Class E basic helix-loop-helix protein 22; trinucleotide repeat containing 20; Trinucleotide repeat-containing gene 20 protein
Gene Aliases: Beta3; BHLHB5; BHLHE22; CAGL85; TNRC20
UniProt ID: (Human) Q8NFJ8
Entrez Gene ID: (Human) 27319
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.