Invitrogen
Manufacturer Code:PA572673
Catalog # PIPA572673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 51-82 amino acids from the N-terminal region of DANRE bhlha9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Class A basic helix-loop-helix protein 9 bHLHa9 Class B basic helix-loop-helix factor 42 bHLHf42 bhlha9 bhlhf42; Class B basic helix-loop-helix factor 42; Orphan1a
Gene Aliases: bhlhf42; wu:fb99e06
Entrez Gene ID: (Zebrafish) 323481
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.