Invitrogen
Manufacturer Code:PA568907
Catalog # PIPA568907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of mouse BHLHA9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHa9; bHLHf42; Class A basic helix-loop-helix protein 9; Class A basic helix-loop-helix protein 9 bHLHa9 Class F basic helix-loop-helix factor 42 bHLHf42 BHLHA9 BHLHF42; Class B basic helix-loop-helix factor 42; novel helix-loop-helix DNA-binding domain containing protein
Gene Aliases: A830053O21Rik; Bhlha9; Bhlhf42; Fingerin
UniProt ID: (Mouse) Q5RJB0
Entrez Gene ID: (Mouse) 320522
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.