Invitrogen
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | 1:1,000 |
Host / Isotype | Rabbit / IgG |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 57-83 amino acids from the Central region of human BHLHA9. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | Lot-Specific |
Storage conditions | -20° C, Avoid Freeze/Thaw Cycles |
RRID | AB_2634542 |
Predicted to react with mouse based on sequence homology.
Putative transcription factor, which may play a role in limb development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.