Novus Biologicals
Manufacturer Code:NBP237997
Catalog # NBP237997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: SKARRMAANVRERKRILDYNEAFNALRRALRHDLGGKRLSKIATLRRAIHRIAALSLVLRASPAPRGPCGHLECHG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic Helix-Loop-Helix Family Member A9 bHLHa9 BHLHF42 Class A Basic Helix-Loop-Helix Protein 9 Class F Basic Helix-Loop-Helix Factor 42 Class II Basic Helix-Loop-Helix Protein; basic helix-loop-helix family, member a9; bHLHa9; bHLHf42; Class A basic helix-loop-helix protein 9; Class F basic helix-loop-helix factor 42; class II basic helix-loop-helix protein; Fingerin
Gene Aliases: BHLHA9; BHLHF42
UniProt ID: (Human) Q7RTU4
Entrez Gene ID: (Human) 727857
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.