Invitrogen
Manufacturer Code:PA568877
Catalog # PIPA568877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the n terminal region of mouse BHE23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix domain containing, class B4; beta-cell E-box transactivating factor 4; BETA4 BHLHB4 bA305P22.3 basic helix-loop-helix domain containing class B 4 class B basic helix-loop-helix protein 4 class E basic helix-loop-helix protein 23; bHLHb4; bHLHe23; Class B basic helix-loop-helix protein 4; Class E basic helix-loop-helix protein 23
Gene Aliases: A930001L02Rik; BETA4; Bhlhb4; Bhlhe23; H20; RGD1559760
UniProt ID: (Mouse) Q8BGW3
Entrez Gene ID: (Rat) 499952, (Mouse) 140489
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.