Novus Biologicals
Manufacturer Code:NB10056094
Catalog # NB10056094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A recombinant protein corresponding to amino acids 1-139 of human BAR was used as immunogen. Human BAR is 450 amino acid protein GenBank no. NP_057645.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BARRNF47RING finger protein 47 bifunctional apoptosis inhibitor bifunctional apoptosis regulator; bifunctional apoptosis inhibitor; Bifunctional apoptosis regulator; RING finger protein 47
Gene Aliases: BAR; BFAR; RNF47
UniProt ID: (Human) Q9NZS9
Entrez Gene ID: (Human) 51283
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.