Novus Biologicals
Manufacturer Code:NBP2865840.1ML
Catalog # NB034208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide corresponding to a region of Mouse BFAR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BARRNF47RING finger protein 47 bifunctional apoptosis inhibitor bifunctional apoptosis regulator; bifunctional apoptosis inhibitor; Bifunctional apoptosis regulator; RING finger protein 47
Gene Aliases: BAR; BFAR; RNF47
UniProt ID: (Human) Q9NZS9
Entrez Gene ID: (Human) 51283
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.