OriGene
Manufacturer Code:TA501979
Catalog # 50-167-3075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H3 |
Immunogen | Full length human recombit protein of human BEST3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bestrophin-3; vitelliform macular dystrophy 2-like 3; Vitelliform macular dystrophy 2-like protein 3; VMD2L3
Gene Aliases: BEST3; VMD2L3
UniProt ID: (Human) Q8N1M1
Entrez Gene ID: (Human) 144453
Molecular Function:
anion channel
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.