Novus Biologicals
Manufacturer Code:NBP237276
Catalog # NBP237276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D9C5 |
Immunogen | Purified recombinant fragment of human BDNF (AA: 19-248) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Abrineurin; abrineurin brain-derived neurotrophic factor MGC34632 neurotrophin; BDNF; BDNF precursor form; Brain-derived neurotrophic factor; neurotrophin; ProBDNF
Gene Aliases: ANON2; BDNF; BULN2
UniProt ID: (Human) P23560
Entrez Gene ID: (Human) 627
Molecular Function:
growth factor
neurotrophic factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.