Proteintech
Manufacturer Code:662921IG150UL
Catalog # 50-173-6724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E5C5 |
Immunogen | BDNF Fusion Protein Ag11329 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Abrineurin; Abrineurin BDNF; BDNF; BDNF precursor form; brain derived neurothrophic factor; brain derived neurotrophic factor; Brain-derived neurotrophic factor; neurotrophin; ProBDNF
Gene Aliases: ANON2; BDNF; BULN2
UniProt ID: (Human) P23560, (Rat) P23363
Entrez Gene ID: (Human) 627, (Rat) 24225
Molecular Function:
growth factor
neurotrophic factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.