Proteintech
Manufacturer Code:282051AP150UL
Catalog # 50-173-5829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BDNF Fusion Protein Ag28276 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abrineurin; Abrineurin BDNF; BDNF; BDNF precursor form; Brain-derived neurotrophic factor; neurotrophin; ProBDNF
Gene Aliases: ANON2; BDNF; BULN2
UniProt ID: (Human) P23560
Entrez Gene ID: (Human) 627
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.