Invitrogen
Manufacturer Code:PA562254
Catalog # PIPA562254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human B2R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B2 bradykinin receptor; B2R; BDKRB2 BK-2 receptor Bradykinin type-2 receptor B2R Bradykinin B2 receptor BRB2 BK-2R BK-2Rs BK2 B2 bradykinin receptor B2bkr BK-2 BKR2 Bradykinin receptor B2; BK-2 receptor
Gene Aliases: B2R; BDKRB2; BK-2; BK2; BKR2; BRB2
UniProt ID: (Human) P30411
Entrez Gene ID: (Human) 624
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.