Abnova
Manufacturer Code:PAB7493
Catalog # 89-133-304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human BDKRB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of BDKRB1.
For Research Use Only
Protein Aliases: B1 bradykinin receptor; B1BKR; B1R; BK-1 receptor; BKB1R; BKR1; BRADYB1; bradykinin B1 receptor; bradykinin receptor 1; bradykinin receptor B2
Gene Aliases: B1BKR; B1R; BDKRB1; BDKRB2; BKB1R; BKR1; BRADYB1
UniProt ID: (Human) P46663
Entrez Gene ID: (Human) 623
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.