Abnova
Manufacturer Code:PAB28617
Catalog # 89-130-129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human BDKRB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant BDKRB1.
For Research Use Only
Protein Aliases: B1 bradykinin receptor; B1BKR B1R BKB1R BKR1 BRADYB1; B1R; BK-1 receptor; bradykinin B1 receptor; bradykinin receptor 1; bradykinin receptor B2
Gene Aliases: B1BKR; B1R; BDKRB1; BDKRB2; BKB1R; BKR1; BRADYB1
UniProt ID: (Human) P46663
Entrez Gene ID: (Human) 623
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.