Invitrogen
Manufacturer Code:PA544760
Catalog # PIPA544760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human BDH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxybutyrate dehydrogenase type 2; 3-hydroxybutyrate dehydrogenase, type 2; dehydrogenase/reductase (SDR family) member 6; Dehydrogenase/reductase SDR family member 6; Oxidoreductase UCPA; R-beta-hydroxybutyrate dehydrogenase; Short chain dehydrogenase/reductase family 15C member 1; short chain dehydrogenase/reductase family 15C, member 1
Gene Aliases: 1810026B04Rik; BDH2; DHRS6; EFA6R; PRO20933; SDR15C1; si:dkey-162b23.2; UCPA-OR; UNQ6308; UNQ6308/PRO20933; zgc:110323
UniProt ID: (Bovine) Q3T046, (Human) Q9BUT1, (Mouse) Q8JZV9, (Rat) D4A1J4, (Zebrafish) Q561X9
Entrez Gene ID: (Rabbit) 100344675, (Bovine) 515321, (Human) 56898, (Pig) 100522692, (Horse) 100068172, (Mouse) 69772, (Guinea pig) 100717546, (Rat) 295458, (Zebrafish) 550507
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.