Abnova
Manufacturer Code:PAB7524
Catalog # 89-133-332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 92-103 of human BDH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of BDH2.
For Research Use Only
Protein Aliases: 3-hydroxybutyrate dehydrogenase type 2; dehydrogenase/reductase (SDR family) member 6; Dehydrogenase/reductase SDR family member 6; DHRS6; EFA6R; FLJ13261; Oxidoreductase UCPA; PRO20933; R-beta-hydroxybutyrate dehydrogenase; SDR15C1; Short chain dehydrogenase/reductase family 15C member 1; short chain dehydrogenase/reductase family 15C, member 1; UCPA-OR; UNQ6308
Gene Aliases: BDH2; DHRS6; EFA6R; PRO20933; SDR15C1; UCPA-OR; UNQ6308; UNQ6308/PRO20933
UniProt ID: (Human) Q9BUT1
Entrez Gene ID: (Human) 56898
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.