Invitrogen
Manufacturer Code:MA515594
Catalog # PIMA515594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A5 |
Immunogen | Purified recombit fragment of human BDH1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (R)-3-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase (heart, mitochondrial); b-hydroxybutyrate dehydrogenase; BDH; BDH SDR9C1 MGC2723 MGC4347 MGC9788 BDH1; D-beta-hydroxybutyrate dehydrogenase, mitochondrial; Short chain dehydrogenase/reductase family 9C member 1; short chain dehydrogenase/reductase family 9C, member 1
Gene Aliases: 2310032J20Rik; AI327223; BDH; BDH1; SDR9C1
UniProt ID: (Human) Q02338, (Mouse) Q80XN0
Entrez Gene ID: (Human) 622, (Mouse) 71911
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.