Novus Biologicals
Manufacturer Code:NBP261707
Catalog # NBP2-61707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E11G12 |
Immunogen | Purified recombinant fragment of human BCR (AA: 139-280) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: ALLD22S11 BCR1 breakpoint cluster region breakpoint cluster region protein CMLEC 2.7.11.1 D22S662 PHLFLJ16453 Renal carcinoma antigen NY-REN-26; BCR/FGFR1 chimera protein; breakpoint cluster region; Breakpoint cluster region protein; FGFR1/BCR chimera protein; Renal carcinoma antigen NY-REN-26
Gene Aliases: ALL; BCR; BCR1; CML; D22S11; D22S662; PHL
UniProt ID: (Human) P11274
Entrez Gene ID: (Human) 613
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.