Novus Biologicals
Manufacturer Code:NB1002665V
Catalog # NB1002665V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C12 |
Immunogen | BCR-ABL peptide |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABL ALL BCR ABL fusion protein bcr/abl BCR/ABL fusion protein Bcr/c abl oncogene protein BCR-ABL fusion protein BCRe18/ABL1e3 fusion protein breakpoint cluster region protein c-ABL CML D22S662 JTK7 p150 PHL; BCR/FGFR1 chimera protein; breakpoint cluster region; Breakpoint cluster region protein; FGFR1/BCR chimera protein; Renal carcinoma antigen NY-REN-26
Gene Aliases: ALL; BCR; BCR1; CML; D22S11; D22S662; PHL
UniProt ID: (Human) P11274
Entrez Gene ID: (Human) 613
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.