Novus Biologicals
Manufacturer Code:NBP246571
Catalog # NBP246571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F6 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 384-668 of human BCOR(NP_060215) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANOP2 BCL6 corepressor BCL-6 corepressor BCL6 co-repressor BCL-6 interacting corepressor BCoR FLJ20285 FLJ38041 KIAA1575MGC71031 MAA2 MCOPS2 MGC131961; BCL-6 corepressor; BCL-6 interacting corepressor; BCoR
Gene Aliases: ANOP2; BCOR; KIAA1575; MAA2; MCOPS2
UniProt ID: (Human) Q6W2J9
Entrez Gene ID: (Human) 54880
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.