Invitrogen
Manufacturer Code:PA5106545
Catalog # PIPA5106545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BCOR corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5830466J11Rik 8430401K06Rik ANOP2 BCL6 corepressor BCL-6 corepressor BCL6 co-repressor BCL6 interacting corepressor BCL-6 interacting corepressor Bcor BcorR D930024N20Rik KIAA1575 MAA2 MCOPS2 mKIAA1575 RGD1562735; BCL-6 corepressor; BCL-6 interacting corepressor; BCoR
Gene Aliases: 5830466J11Rik; 8430401K06Rik; ANOP2; BCOR; BcorR; D930024N20Rik; KIAA1575; MAA2; MCOPS2; mKIAA1575
UniProt ID: (Human) Q6W2J9, (Mouse) Q8CGN4
Entrez Gene ID: (Human) 54880, (Mouse) 71458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.