Novus Biologicals
Manufacturer Code:NB10087005
Catalog # NB10087005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1705 and 1755 of human BCL6 co-repressor using the numbering given in entry EAW59426.1 (GeneID 54880). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANOP2 BCL6 corepressor BCL-6 corepressor BCL6 co-repressor BCL-6 interacting corepressor BCoR FLJ20285 FLJ38041 KIAA1575MGC71031 MAA2 MCOPS2 MGC131961; BCL-6 corepressor; BCL-6 interacting corepressor; BCoR
Gene Aliases: ANOP2; BCOR; KIAA1575; MAA2; MCOPS2
UniProt ID: (Human) Q6W2J9
Entrez Gene ID: (Human) 54880
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.