Novus Biologicals
Manufacturer Code:NBP298770AF488
Catalog # NB055332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human BCOR. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANOP2 BCL6 corepressor BCL-6 corepressor BCL6 co-repressor BCL-6 interacting corepressor BCoR FLJ20285 FLJ38041 KIAA1575MGC71031 MAA2 MCOPS2 MGC131961; BCL-6 corepressor; BCL-6 interacting corepressor; BCoR
Gene Aliases: ANOP2; BCOR; KIAA1575; MAA2; MCOPS2
UniProt ID: (Human) Q6W2J9
Entrez Gene ID: (Human) 54880
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.