Invitrogen
Manufacturer Code:PA5104323
Catalog # PIPA5104323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BCO2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ab2-079 bb-carotene-9'10'-dioxygenase Bcdo2 Bcmo2 Bco2 B-DIOX-II betabeta-carotene 9'10'-dioxygenase variant 1 betabeta-carotene 9'10'-dioxygenase variant 2 betabeta-carotene 9'10'-dioxygenase variant 3 Betabeta-carotene 9'10'-oxygenase beta-carotene 9' 10'-dioxygenase 2 beta-carotene 9'10' oxygenase beta-carotene 9'10'-dioxygenase 2 beta-carotene dioxygenase 2 beta-carotene oxygenase 2 beta-diox-II carotenoid 9'10' monooxygenase II carotenoid 9'10' monoxygenase II carotenoid-9'10'-cleaving dioxygenase CMO2; b,b-carotene-9',10'-dioxygenase; B-diox-II; beta,beta-carotene 9',10'-dioxygenase variant 1; beta,beta-carotene 9',10'-dioxygenase variant 2; beta,beta-carotene 9',10'-dioxygenase variant 3; Beta,beta-carotene 9',10'-oxygenase; beta-carotene 9', 10'-dioxygenase 2; beta-carotene 9',10' oxygenase; beta-carotene 9',10'-dioxygenase 2; Beta-carotene dioxygenase 2; beta-diox-II; carotenoid 9',10' monooxygenase II; carotenoid 9',10' monoxygenase II; carotenoid-9',10'-cleaving dioxygenase
Gene Aliases: Ab2-079; B-DIOX-II; BCDO2; Bcmo2; BCO2; CMO2
UniProt ID: (Human) Q9BYV7, (Mouse) Q99NF1
Entrez Gene ID: (Human) 83875, (Rat) 315644, (Mouse) 170752
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.