Abnova
Manufacturer Code:PAB23586
Catalog # 89-125-291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human BCO2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant BCO2.
For Research Use Only
Protein Aliases: b,b-carotene-9',10'-dioxygenase; B-DIOX-II; BCDO2; beta,beta-carotene 9',10'-dioxygenase variant 1; beta,beta-carotene 9',10'-dioxygenase variant 2; beta,beta-carotene 9',10'-dioxygenase variant 3; Beta,beta-carotene 9',10'-oxygenase; beta-carotene 9',10' oxygenase; Beta-carotene dioxygenase 2; carotenoid-9',10'-cleaving dioxygenase; FLJ34464
Gene Aliases: B-DIOX-II; BCDO2; BCO2
UniProt ID: (Human) Q9BYV7
Entrez Gene ID: (Human) 83875
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.