Invitrogen
Manufacturer Code:PA5114226
Catalog # PIPA5114226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: VEKGKTKYVI FKIPATVPEG KKQGKSPWKH TEVFCSIPSR SLLSPSYYHS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bcdo Bcdo1 BCMO Bcmo1 BCO BCO1 Betabeta-carotene 1515&prime-dioxygenase betabeta-carotene 1515&prime-monooxygenase beta-carotene 15 15&prime-dioxygenase beta-carotene 15 15-dioxygenase beta-carotene 15 15&prime-dioxygenase beta-carotene 15 15-dioxygenase beta-carotene 15 15&prime-dioxygenase 1 beta-carotene 1515&prime dioxygenase beta-carotene 1515&prime-monooxygenase beta-carotene 1515&prime-monooxygenase 1 beta-carotene cleavage enzyme Beta-carotene dioxygenase 1 Beta-carotene oxygenase 1 beta-CD betaCMOOX carotenoid-1515&prime-oxygenase CMO1 Cmoi; Beta,beta-carotene 15,15'-dioxygenase; beta,beta-carotene 15,15'-monooxygenase; beta-carotene 15, 15'-dioxygenase 1; beta-carotene 15,15'-monooxygenase 1; Beta-carotene dioxygenase 1; Beta-carotene oxygenase 1
Gene Aliases: BCDO; BCDO1; BCMO; BCMO1; BCO; BCO1
UniProt ID: (Human) Q9HAY6
Entrez Gene ID: (Human) 53630
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.