Novus Biologicals
Manufacturer Code:NBP292267100UL
Catalog # NB026425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 268-547 of human BCMO1 (NP_059125.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCDO1 BCDOEC 1.14.99.36 BCMO BCO BCO1 beta beta-carotene 15 15&prime-dioxygenase 1 beta-carotene 1515&prime-monooxygenase beta-carotene 1515&prime-monooxygenase 1 Beta-carotene dioxygenase 1 FLJ10730; Beta,beta-carotene 15,15'-dioxygenase; beta,beta-carotene 15,15'-monooxygenase; beta-carotene 15, 15'-dioxygenase 1; beta-carotene 15,15'-monooxygenase 1; Beta-carotene dioxygenase 1; Beta-carotene oxygenase 1
Gene Aliases: BCDO; BCDO1; BCMO; BCMO1; BCO; BCO1
UniProt ID: (Human) Q9HAY6
Entrez Gene ID: (Human) 53630
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.