Proteintech
Manufacturer Code:233081AP150UL
Catalog # 50-173-4044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BCMO1 Fusion Protein Ag19732 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCMO1 Beta carotene dioxygenase 1; Beta,beta-carotene 15,15'-dioxygenase; beta,beta-carotene 15,15'-monooxygenase; beta-carotene 15, 15'-dioxygenase 1; beta-carotene 15,15' dioxygenase; beta-carotene 15,15'-monooxygenase; beta-carotene 15,15'-monooxygenase 1; beta-carotene cleavage enzyme; Beta-carotene dioxygenase 1; Beta-carotene oxygenase 1; carotenoid-15,15'-oxygenase
Gene Aliases: BCDO; BCDO1; BCMO; BCMO1; BCO; BCO1; beta-CD; betaCMOOX; CMO1; Cmoi
UniProt ID: (Human) Q9HAY6, (Mouse) Q9JJS6
Entrez Gene ID: (Human) 53630, (Mouse) 63857
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.