Invitrogen
Manufacturer Code:14988737
Catalog # 14-988-737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GI191E |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B cell CLL/lymphoma 6 B cell leukemia/lymphoma 6 B-cell CLL/lymphoma 6 B-cell CLL/lymphoma 6 (zinc finger protein 51) B-cell leukemia/lymphoma 5 B-cell leukemia/lymphoma 6 B-cell lymphoma 5 protein B-cell lymphoma 6 protein B-cell lymphoma 6 protein homolog B-cell lymphoma 6 protein transcript BCL5 BCL-5 BCL6 BCL-6 BCL6 transcription repressor BCL6A cys-his2 zinc finger transcription factor LAZ3 LAZ-3 protein lymphoma-associated zinc finger gene on chromosome 3 protein LAZ-3 ZBTB27 Zinc finger and BTB domain-containing protein 27 zinc finger protein 51 zinc finger transcription factor BCL6S ZNF51; B-cell leukemia/lymphoma 5; B-cell leukemia/lymphoma 6; B-cell lymphoma 5 protein; B-cell lymphoma 6 protein; B-cell lymphoma 6 protein homolog; B-cell lymphoma 6 protein transcript; BCL-5; BCL-6; cys-his2 zinc finger transcription factor; lymphoma-associated zinc finger gene on chromosome 3; Protein LAZ-3; Zinc finger and BTB domain-containing protein 27; Zinc finger protein 51; zinc finger transcription factor BCL6S
Gene Aliases: Bcl-6; BCL5; BCL6; BCL6A; LAZ3; ZBTB27; ZNF51
UniProt ID: (Human) P41182, (Mouse) P41183
Entrez Gene ID: (Human) 604, (Mouse) 12053
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.