Invitrogen
Manufacturer Code:PA527390
Catalog # PIPA527390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 160 and 415 of Human BCL6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell leukemia/lymphoma 5; B-cell leukemia/lymphoma 6; B-cell lymphoma 5 protein; B-cell lymphoma 6 protein; B-cell lymphoma 6 protein homolog; B-cell lymphoma 6 protein transcript; BCL-5; BCL-6; BCL5 BCL6A LAZ3 ZBTB27 ZNF51 B-cell lymphoma 5 protein B-cell lymphoma 6 protein B-cell lymphoma 6 protein transcript BCL-5 BCL-6 cys-his2 zinc finger transcription factor lymphoma-associated zinc finger gene on chromosome 3 protein LAZ-3 zinc finger and BTB domain-containing protein; cys-his2 zinc finger transcription factor; lymphoma-associated zinc finger gene on chromosome 3; Protein LAZ-3; Zinc finger and BTB domain-containing protein 27; Zinc finger protein 51; zinc finger transcription factor BCL6S
Gene Aliases: Bcl-6; BCL5; BCL6; BCL6A; LAZ3; ZBTB27; ZNF51
UniProt ID: (Human) P41182, (Mouse) P41183
Entrez Gene ID: (Human) 604, (Mouse) 12053
Molecular Function: DNA binding protein centromere DNA-binding protein nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.