Abnova
Manufacturer Code:PAB8573
Catalog # 89-133-758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 56 of human BCL2L1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of BCL2L1.
For Research Use Only
Protein Aliases: Apoptosis regulator Bcl-X; Bcl-2-like protein 1; Bcl-X; bcl-xL; BCL-XL/S; bcl-xS; Bcl2-L-1; BCL2-like 1; BCL2L; BCLX; DKFZp781P2092; protein phosphatase 1, regulatory subunit 52
Gene Aliases: Bcl-X; BCL-XL/S; BCL2L; BCL2L1; BCLX; PPP1R52
UniProt ID: (Human) Q07817
Entrez Gene ID: (Human) 598
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.