Invitrogen
Manufacturer Code:MA531434
Catalog # PIMA531434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL6426 |
Immunogen | Synthetic peptide corresponding to Human BCL11B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell CLL/lymphoma 11B; B-cell CLL/lymphoma 11B (zinc finger protein); B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein; B-cell leukemia/lymphoma 11B; B-cell lymphoma/leukaemia 11B; B-cell lymphoma/leukemia 11B; BCL-11B; BCL11B/TRDC fusion; COUP-TF interacting protein 2; COUP-TF-interacting protein 2; CTIP-2 CTIP2 hRIT1-alpha ZNF856B; hRit1; mRit1; Radiation-induced tumor suppressor gene 1 protein; zinc finger protein hRit1 alpha
Gene Aliases: 9130430L19Rik; AI604821; ATL1; ATL1-alpha; ATL1-beta; ATL1-delta; ATL1-gamma; B630002E05Rik; BCL-11B; BCL11B; CTIP-2; CTIP2; hRIT1-alpha; RIT1; ZNF856B
UniProt ID: (Human) Q9C0K0, (Mouse) Q99PV8
Entrez Gene ID: (Human) 64919, (Mouse) 58208
Molecular Function:
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.