Invitrogen
Manufacturer Code:MA534678
Catalog # PIMA534678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB35-68 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2810047E18Rik B-cell CLL/lymphoma 11A B-cell lymphoma/leukemia 11A Bcl11a BCL-11A BCL11A-L BCL11a-M BCL11A-S BCL11A-XL BOS_11500 COUP-TF-interacting protein 1 CTIP1 D930021L15Rik Ecotropic viral integration site 9 protein Ecotropic viral integration site 9 protein homolog EVI9 EVI-9 Evi9a Evi9b Evi9c FLJ10173 FLJ34997 HBFQTL5 Kiaa1809 mKIAA1809 Zinc finger protein 856 ZNF856; B-cell CLL/lymphoma 11A; B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2; B-cell lymphoma/leukemia 11A; BCL-11A; BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1; C2H2-type zinc finger protein; COUP-TF-interacting protein 1; ecotropic viral integration site 9 homolog; Ecotropic viral integration site 9 protein homolog; EVI-9; Zinc finger protein 856
Gene Aliases: BCL11A; BCL11A-L; BCL11a-M; BCL11A-S; BCL11A-XL; CTIP1; EVI9; HBFQTL5; KIAA1809; ZNF856
UniProt ID: (Human) Q9H165
Entrez Gene ID: (Human) 53335
Molecular Function: nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.