Invitrogen
Manufacturer Code:MA535783
Catalog # PIMA535783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1486 |
Immunogen | A synthesized peptide derived from human CTIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2810047E18Rik B-cell CLL/lymphoma 11A B-cell lymphoma/leukemia 11A Bcl11a BCL-11A BCL11A-L BCL11a-M BCL11A-S BCL11A-XL BOS_11500 COUP-TF-interacting protein 1 CTIP1 D930021L15Rik Ecotropic viral integration site 9 protein Ecotropic viral integration site 9 protein homolog EVI9 EVI-9 Evi9a Evi9b Evi9c FLJ10173 FLJ34997 HBFQTL5 Kiaa1809 mKIAA1809 Zinc finger protein 856 ZNF856; B-cell lymphoma/leukemia 11A
Entrez Gene ID: (Rat) 305589
Molecular Function: nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.