Invitrogen
Manufacturer Code:PA568505
Catalog # PIPA568505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse BCL11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B cell CLL/lymphoma 11A; B cell CLL/lymphoma 11A (zinc finger protein); B-cell CLL/lymphoma 11A; B-cell CLL/lymphoma 11A (zinc finger protein); B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2; B-cell lymphoma/leukemia 11A; BCL-11A; BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1; BCL11A, BAF complex component; BCL11A-L BCL11A-S BCL11A-XL CTIP1 EVI9 HBFQTL5 ZNF856 B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2 B-cell lymphoma/leukemia 11A BCL-11A BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1 C2H2-type zinc finger protein COUP-TF-interacting protein 1 EVI-9 ecotropic viral integration site; C2H2-type zinc finger protein; COUP-TF interacting protein 1; COUP-TF-interacting protein 1; ecotropic viral integration site 9 homolog; Ecotropic viral integration site 9 protein; Ecotropic viral integration site 9 protein homolog; EVI-9; myeloid leukemia; Zinc finger protein 856
Gene Aliases: 2810047E18Rik; BCL-11A; BCL11A; BCL11A-L; BCL11a-M; BCL11A-S; BCL11A-XL; CTIP1; D930021L15Rik; EVI9; Evi9a; Evi9b; Evi9c; HBFQTL5; KIAA1809; mKIAA1809; ZNF856
UniProt ID: (Human) Q9H165, (Mouse) Q9QYE3
Entrez Gene ID: (Dog) 481381, (Bovine) 538680, (Horse) 100065364, (Human) 53335, (Rat) 305589, (Guinea pig) 100734801, (Mouse) 14025
Molecular Function: nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.