Invitrogen
Manufacturer Code:PA129817
Catalog # PIPA129817
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to amino acids 2 - 13 and 107 - 120 of Human Bcl3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL-3; BCL4 D19S37; chronic lymphatic leukemia protein; Proto-oncogene BCL3
Gene Aliases: AI528691; Bcl-3; BCL3; BCL4; D19S37
UniProt ID: (Human) P20749, (Mouse) Q9Z2F6
Entrez Gene ID: (Human) 602, (Mouse) 12051, (Rat) 680611
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.