Novus Biologicals
Manufacturer Code:NBP147664
Catalog # NBP147664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI11C9 |
Immunogen | Full length human recombinant protein of human BCKDK (NP_005872) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: [3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial; BCKD-kinase; BCKDHKIN; BCKDHKIN branched chain alpha-ketoacid dehydrogenase kinase branched chain ketoacid dehydrogenase kinase EC 2.7.11 EC 2.7.11.4 mitochondrial; branched chain alpha-ketoacid dehydrogenase kinase; Branched-chain alpha-ketoacid dehydrogenase kinase
Gene Aliases: BCKDK; BCKDKD; BDK
UniProt ID: (Human) O14874
Entrez Gene ID: (Human) 10295
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.