Invitrogen
Manufacturer Code:PA528050
Catalog # PIPA528050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 77 and 367 of Human BCKDK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial; BCKD-kinase; BCKD-kinase BCKDHKIN &lsqb3-methyl-2-oxobutanoate dehydrogenase &lsqblipoamide]] kinase mitochondrial branched chain alpha-ketoacid dehydrogenase kinase branched-chain alpha-ketoacid dehydrogenase kinase; BCKDHKIN; branched chain alpha-ketoacid dehydrogenase kinase; Branched-chain alpha-ketoacid dehydrogenase kinase
Gene Aliases: AI327402; BCKDK; BCKDKD; BDK
UniProt ID: (Human) O14874, (Mouse) O55028
Entrez Gene ID: (Human) 10295, (Mouse) 12041
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.