Invitrogen
Manufacturer Code:PA5104476
Catalog # PIPA5104476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BCKD corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase mitochondrial AI327402 BCKDHKIN Bckdk BCKDKD BCKD-kinase BDK branched chain alpha-ketoacid dehydrogenase kinase branched chain ketoacid dehydrogenase kinase branched-chain alpha-ketoacid dehydrogenase kinase EC 2.7.11.4; [3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial; BCKD-kinase; BCKDHKIN; branched chain alpha-ketoacid dehydrogenase kinase; Branched-chain alpha-ketoacid dehydrogenase kinase
Gene Aliases: AI327402; BCKDK; BCKDKD; BDK
UniProt ID: (Human) O14874, (Rat) Q00972, (Mouse) O55028
Entrez Gene ID: (Human) 10295, (Rat) 29603, (Mouse) 12041
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.