Invitrogen
Manufacturer Code:PA524166
Catalog # PIPA524166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 41-70 amino acids from the N-terminal region of human BCKDHB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoisovalerate dehydrogenase subunit beta, mitochondrial; BCKDE1B; branched chain alpha-ketoacid dehydrogenase E1-beta subunit; branched chain keto acid dehydrogenase E1, beta polypeptide; Branched-chain alpha-keto acid dehydrogenase E1 component beta chain; E1B dJ279A18.1 2-oxoisovalerate dehydrogenase beta subunit 2-oxoisovalerate dehydrogenase subunit beta mitochondrial BCKDE1B BCKDH E1-beta E1b-beta subunit of the branched-chain complex branched chain alpha-ketoacid dehydrogenase E1-beta subunit branched-chain alpha-ke; E1b-beta subunit of the branched-chain complex; testis secretory sperm-binding protein Li 240mP
Gene Aliases: BCKDE1B; BCKDH E1-beta; BCKDHB; E1B
UniProt ID: (Human) P21953
Entrez Gene ID: (Human) 594
Molecular Function:
dehydrogenase
lyase
oxidoreductase
transferase
transketolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.