Invitrogen
Manufacturer Code:PA576434
Catalog # PIPA576434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human BCKDHA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoisovalerate dehydrogenase (lipoamide); 2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial; BCKAD E1[a]; BCKDE1A; BCKDE1A BCKDH E1-alpha BCKDHA; BCKDH E1-alpha; branched chain alpha-ketoacid dehydrogenase subunit E1 alpha; branched chain keto acid dehydrogenase E1 alpha protein; branched chain keto acid dehydrogenase subunit E1, alpha polypeptide; Branched-chain alpha-keto acid dehydrogenase E1 component alpha chain
Gene Aliases: BCKDA; BCKDE1A; BCKDHA; E1a; MSU; MSUD1; OVD1A
UniProt ID: (Human) Q16472, (Rat) P11960
Entrez Gene ID: (Human) 593, (Rat) 25244, (Mouse) 12039
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.