Invitrogen
Manufacturer Code:PA5114897
Catalog # PIPA5114897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BCKDHA corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoisovalerate dehydrogenase (lipoamide); 2-oxoisovalerate dehydrogenase (lipoamide) 2-oxoisovalerate dehydrogenase subunit alpha mitochondrial BCKAD E1[a] BCKDA BCKDE1A BCKDH E1-alpha Bckdha branched chain alpha-ketoacid dehydrogenase subunit E1 alpha branched chain keto acid dehydrogenase E1 alpha protein branched chain keto acid dehydrogenase E1 alpha polypeptide branched chain keto acid dehydrogenase subunit E1 alpha polypeptide branched chain ketoacid dehydrogenase E1 alpha polypeptide branched-chain alpha-keto acid dehydrogenase E1 component alpha chain E1a MSU MSUD1 OVD1A; 2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial; BCKAD E1[a]; BCKDE1A; BCKDH E1-alpha; branched chain alpha-ketoacid dehydrogenase subunit E1 alpha; branched chain keto acid dehydrogenase E1 alpha protein; branched chain keto acid dehydrogenase subunit E1, alpha polypeptide; Branched-chain alpha-keto acid dehydrogenase E1 component alpha chain
Gene Aliases: BCKDA; BCKDE1A; BCKDHA; E1a; MSU; MSUD1; OVD1A
UniProt ID: (Human) Q16472, (Rat) P11960
Entrez Gene ID: (Human) 593, (Rat) 25244, (Mouse) 12039
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.