Novus Biologicals
Manufacturer Code:NBP297503V
Catalog # NB097319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human BCKDHA fragment. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoisovalerate dehydrogenase (lipoamide); 2-oxoisovalerate dehydrogenase subunit alpha mitochondrial BCKDE1A BCKDH E1-alpha branched chain keto acid dehydrogenase E1 alpha polypeptide branched chain keto acid dehydrogenase E1 alpha polypeptide (maple syrup urinedisease) Branched-chain alpha-keto acid dehydrogenase E1 component alpha chain EC 1.2.4.42-oxoisovalerate dehydrogenase (lipoamide) FLJ45695 MSU MSUD1 OVD1A; 2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial; BCKDE1A; BCKDH E1-alpha; branched chain keto acid dehydrogenase E1 alpha protein; Branched-chain alpha-keto acid dehydrogenase E1 component alpha chain
Gene Aliases: BCKDE1A; BCKDHA; MSU; MSUD1; OVD1A
UniProt ID: (Human) Q16472
Entrez Gene ID: (Human) 593
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.