Invitrogen
Manufacturer Code:PA577211
Catalog # PIPA577211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human BCAT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCAT(m); branched chain amino-acid transaminase 2, mitochondrial; branched chain aminotransferase 2, mitochondrial; Branched-chain-amino-acid aminotransferase mitochondrial BCAT(m) Placental protein 18 PP18 BCAT2 BCATM BCT2 ECA40; Branched-chain-amino-acid aminotransferase, mitochondrial; heart branched chain aminotransferase precursor encoding mitochondrial protein; Placental protein 18; PP18
Gene Aliases: BCAM; Bcat-2; BCAT2; BCATM; BCT2; ECA40; mBcat; PP18
UniProt ID: (Human) O15382, (Mouse) O35855, (Rat) O35854
Entrez Gene ID: (Human) 587, (Mouse) 12036, (Rat) 64203
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.