Invitrogen
Manufacturer Code:PA569403
Catalog # PIPA569403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse BCAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCAT(c); BCATC BCT1 ECA39 MECA39 PNAS121 PP18 branched chain aminotransferase 1 cytosolic branched-chain-amino-acid aminotransferase cytosolic placental protein 18; branched chain amino-acid transaminase 1, cytosolic; branched chain aminotransferase 1, cytosolic; Branched-chain-amino-acid aminotransferase, cytosolic; cytosolic branched-chain amino acid aminotransferase; placental protein 18; Protein ECA39
Gene Aliases: BCAT1; BCATC; BCT1; ECA39; MECA39; PNAS121; PP18
UniProt ID: (Human) P54687, (Sheep) Q9GKM4, (Mouse) P24288, (Rat) P54690
Entrez Gene ID: (Horse) 100068713, (Human) 586, (Sheep) 443485, (Bovine) 505926, (Dog) 486633, (Pig) 100518784, (Rabbit) 100337981, (Guinea pig) 100722166, (Mouse) 12035, (Rat) 29592
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.