OriGene
Manufacturer Code:CF502614
Catalog # 50-166-1490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A5 |
Immunogen | Full length human recombit protein of human BCAR1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCAR1, Cas family scaffold protein; BCAR1, Cas family scaffolding protein; breast cancer anti-estrogen resistance 1; Breast cancer anti-estrogen resistance protein 1; CAS CAS1 CASS1 CRKAS P130Cas; Cas scaffolding protein family member 1; CRK-associated substrate; Crk-associated substrate p130Cas; p130 Cas; p130cas; v-crk-associated tyrosine kinase substrate
Gene Aliases: AI385681; BCAR1; CAS; CAS1; CASS1; CRKAS; P130CAS
UniProt ID: (Human) P56945, (Mouse) Q61140, (Rat) Q63767
Entrez Gene ID: (Human) 9564, (Dog) 479648, (Mouse) 12927, (Rat) 25414
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.